missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes
GREENER_CHOICE

Catalog No. 055291A
Change view
Click to view available options
Capacity (Metric):
10 mL
28 mL
30 mL
50 mL
Length (Metric):
80.5 mm
93.5 mm
94.0 mm
105.9 mm
Diameter (Metric):
16.1 mm
25.8 mm
28.8 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Length (Metric) Diameter (Metric)
055291A 10 mL 80.5 mm 16.1 mm
055291B 28 mL 94.0 mm 25.8 mm
055291C 30 mL 93.5 mm 25.8 mm
055291D 50 mL 105.9 mm 28.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Must be filled to 80% of total capacity when centrifuging
Catalog No. 055291A Supplier Thermo Scientific™ Supplier No. 31190010
Only null left
Must be filled to 80% of total capacity when centrifuging

View contents of these contact-clear, high-speed PPCO centrifuge tubes easily. The tubes also feature excellent mechanical strength.

Thermo Scientific™ Nalgene™ Oak Ridge High-Speed PPCO Centrifuge Tubes are chemically compatible with a wider variety of lab reagents than polycarbonate centrifuge tubes and are repeately autoclavable for extended performance.

Features:

  • Greater chemical resistance than polycarbonate
  • Can be autoclaved repeatedly—last longer than polycarbonate tubes under conditions of repeated autoclaving
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges; 3119-0050 can be used up to 100,605 x g if equipped with a sealing cap
  • Come with polypropylene linerless closures—easy to clean and provides leakproof protection under normal conditions up to 10,000 x g
  • For leakproof assurance at speeds >10,000 x g use with sealing cap assembly (sold separately)
  • Leakproof† construction ensures more secure storage of samples

Ordering Information:

Optional sealing-cap assemblies available for selected sizes. Use DS3132-0020 (for 26 and 30mL tube) and DS3132-0024 (for 50mL Tube). Actual brim capacity of 3119-0050 is 42mL.

Specifications

Closure Material Polypropylene
Capacity (Metric) 10 mL
Material Polypropylene Copolymer
No. per Case 100
No. per Pack 10
Sterility Non-sterile
Autoclavable Autoclavable
Shape Round
Length (Metric) 80.5 mm
Bottom Shape Round
Closure Type Screw Cap
Writing Area No
Diameter (Metric) 16.1 mm
Graduated No
Product Type Centrifuge Tube
Max. RCF 10,000 x g
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.