Learn More
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes w/Sealing Cap
View contents easily using these strong, transparent polycarbonate centrifuge tubes, available with sealing caps for use up to 50,000 x g.
Supplier: Thermo Scientific™ 31380016
Alert:
Autoclaving repeatedly will shorten the life of polycarbonate tubes. Tubes must be filled to at least 80% capacity for proper performance.
Description
Easily view contents using strong, transparent Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Polycarbonate Centrifuge Tubes, equipped with sealing caps for leakproof† performance up to 50,000 x g.
Features:- Withstand temperatures from -135° to +135°C (-211° to +275°F)
- Round bottom to fit high speed rotors
- Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
- Come with PP sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
- Autoclavable; transparent
Ordering Information:
Sealing caps also sold separately for 30 and 50mL sizes.
Specifications
| Polypropylene, red rubber | |
| Polycarbonate | |
| 10 | |
| Autoclavable | |
| 107.2 mm | |
| Screw Cap | |
| 18.2 mm | |
| Centrifuge Tube |
| 16 mL | |
| 50 | |
| Non-sterile | |
| Round | |
| Round | |
| No | |
| No | |
| 50,000 x g |