missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Microcentrifuge Tube Storage Boxes
GREENER_CHOICE

Catalog No. 1535058
Change view
Click to view available options
Array:
8 x 8
9 x 9
For Use With (Equipment):
0.2mLtubes
0.5mL tubes
1.5mL tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array For Use With (Equipment)
1535058 8 x 8 0.2mLtubes
1535059 9 x 9 0.5mL tubes
1535060 8 x 8 1.5mL tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1535058 Supplier Thermo Scientific™ Supplier No. 50555002

Safely store samples using these microcentrifuge tube storage boxes.

Safely store samples using Thermo Scientific™ Nalgene™ Microcentrifuge Tube Storage Boxes. High-temperature polyurethane foam insert securely holds 64 or 81 microcentrifuge tubes sized 0.2, 0.5 or 1.5mL.

  • Bottom constructed of white polycarbonate
  • Transparent polycarbonate lid has printed grid for inventory purposes
  • Store from -211° to +250°F (-135° to +121°C)
  • Lid accepts marker for box and tube identification and is keyed to orient front and back of box
  • L x W x H: 5.25 x 5.25 x 2 in. (13 x 13 x 5cm)

Note:
Not for use in liquid phase of liquid nitrogen.

Specifications

Color White
Holds Microtubes (0.2 mL)
Material Polyurethane Foam, Polycarbonate
Positions 64
Shape Square
For Use With (Equipment) 0.2mLtubes
No. per Case 24
No. per Pack 4
Product Type Microcentrifuge Tube Storage Box
Array 8 x 8