Learn More
Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
$572.70 - $673.46
| Catalog Number | Mfr. No. | Holds | For Use With (Equipment) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Holds | For Use With (Equipment) | Price | Quantity & Availability | ||||
|
1167668
|
Thermo Scientific™
HR509X20A 70 |
Cryogenic Box, 81-Ampule | For: Locator 4 Plus |
Each for $673.46
|
|
||||
|
1167667
|
Thermo Scientific™
HR509X19A70 |
Cryogenic Box, 81-Ampule | For Locator Jr. Plus |
Each for $572.70
|
|
||||
Description
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
- Designed for long-term storage
- Features an indexed frame and box organizational system
- Unique hanging rack promotes easy sample retrieval
- Suited for both manual and computerized inventory record-keeping methods
Specifications
| Cryogenic Box, 81-Ampule | |
| Locator™ Plus Cryogenic Rack |