missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closure for Continuous-Thread Vials: Nonsterile
GREENER_CHOICE

Catalog No. 03315
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03315 White
14178179 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03315 Supplier Thermo Scientific™ Supplier No. 3121580021

Eliminate the need for a rubber stopper while maintaining a leakproof seal with these lined, HDPE serum vial closures, for use with continuous thread PETG serum vials.

Eliminate the need for a rubber stopper, yet maintain a leakproof† seal with Thermo Scientific™ Nalgene™ Lined HDPE Serum Vial Closures for Continuous-Thread Vials. These nonsterile, high density polyethylene closures are for use with continuous-thread PETG serum vials.

Features:

  • Three-ply co-extruded liner (LDPE core between two facings of solid HDPE)
  • For use with nonsterile Nalgene PETG serum vials

Ordering Information:

Serum vial closures are bulk packed 2,000 per case.

Specifications

Closure Color White
Description Serum vial closures
Material (Liner) Three-ply co-extruded liner (LDPE core between two facing of solid HDPE)
Color White
Diameter (Metric) Outer 22.1 mm
Material HDPE
Sterility Non-sterile
No. per Case 2000
No. per Pack 2000
Product Line Nalgene
Closure Material HDPE
Closure Type Screw Cap
Quantity 2000 Cs.
Closure Size 20-415
For Use With (Equipment) Nonsterile Nalgene PETG Serum Vials
Certifications/Compliance ISO 13485:2003
Height (Metric) 14.2 mm
Product Type Serum Vial Closure
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.