Learn More
Thermo Scientific™ Nalgene™ LDPE Sample Bags
Keep dry samples dry and moist samples moist with these LDPE sample bags, ideal for small lab items, botanical samples and other specimens.
Supplier: Thermo Scientific™ 62550508
Alert:
Not recommended for storing liquids.
Description
Keep dry samples dry and moist samples moist with Thermo Scientific™ Nalgene™ LDPE Sample Bags. Ideal for small lab items, botanical samples and other specimens, these heavy-duty, double-track, low-density polyethylene zipper bags lock securely, but are not recommended for storing liquids.
- Low-density polyethylene is clear for easy identification of contents
- 4mil thick walls are strong to protect your samples
- Bags are waterproof to keep contents dry
- Transparent
Chemical Identifiers
| Heavy-duty double-track zipper bag ideal for small lab items, botanical samples and other specimens | |
| Transparent | |
| LDPE |
Specifications
| Heavy-duty double-track zipper bag ideal for small lab items, botanical samples and other specimens | |
| LDPE | |
| 5 in. | |
| 12.7 cm |
| Transparent | |
| 8 in. | |
| 20.3 cm |