missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE, Round Carboy with Spigot

For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White

Supplier:  Thermo Scientific™ 23180010

 View more versions of this product

Catalog No. 02-963AA

Only null left
Explore available promotions

Description

Description

Thermo Scientific™ Nalgene Round LDPE Carboys are the icon of lab water dispensing and storage. Place one at every sink and every work station to enhance your productivity. Carboys have a spigot boss molded in once piece with the carboy for convenience, strength and leak resistance. Suitable for collecting and dispensing distilled water, acids and reagents. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

  • Molded in graduations in 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles make lifting comfortable for four largest sizes
  • Molded in spigot boss provides strength and leak resistance
  • Leakproof†

Ordering Information:

Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.

Includes:

Polypropylene spigot and screw closure with TPE gasket † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Specifications

Not Autoclavable
4 L
38-430
Round Carboys w/Spigot
154 mm
Molded-in graduations in 1 gal. and 5 L increments
Integral shoulder handles, Molded-in spigot boss
6/Cs.
1 gal.
Polypropylene
White
26 mm
Collect and dispense distilled water, acids and reagents
335 mm
LDPE
Carboy with Spigot
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions