missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE, Round Carboy with Spigot
GREENER_CHOICE

Catalog No. 02963A
Change view
Click to view available options
Capacity (Metric):
4 L
10 L
20 L
25 L
50 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02963A 10 L
02963AA 4 L
02963B 25 L
02963BB 20 L
02963C 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02963A Supplier Thermo Scientific™ Supplier No. 23180020
Get Extra Thermo Scientific Products with Purchase
View all promotions

For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White

Thermo Scientific™ Nalgene Round LDPE Carboys are the icon of lab water dispensing and storage. Place one at every sink and every work station to enhance your productivity. Carboys have a spigot boss molded in once piece with the carboy for convenience, strength and leak resistance. Suitable for collecting and dispensing distilled water, acids and reagents. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.

  • Molded in graduations in 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles make lifting comfortable for the four largest sizes
  • Molded-in spigot boss provides strength and leak resistance
  • Leakproof†

Ordering Information:

Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.

Includes:

Polypropylene spigot and screw closure with TPE gasket † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Material Polypropylene
Closure Size 83B
Color White (Opaque)
Description Round Carboys w/Spigot
Diameter (Metric) Inner Neck 64 mm
Diameter (Metric) Outer 250 mm
For Use With (Application) Collect and dispense distilled water, acids and reagents
Graduations Molded-in graduations in 1 gal. and 5 L increments
Height (Metric) With Closure 389 mm
Includes Integral shoulder handles, Molded-in spigot boss
Material LDPE
Product Type Carboy with Spigot
Show More Show Less