missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE, Round Carboy
GREENER_CHOICE

Catalog No. 02961D
Change view
Click to view available options
Capacity (Metric):
10 L
15 L
20 L
25 L
50 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02961D 10 L
02961E 15 L
02961A 20 L
02961B 25 L
02961C 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02961D Supplier Thermo Scientific™ Supplier No. 22100020

For Use With: Store and transport reagents; Closure Material: Polypropylene; Color: White

Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.

  • Molded-in graduations of 5 L and 1 gal. increments
  • Leakproof†

Includes:White polypropylene screw cap and 83B closure

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Closure Material Polypropylene
Closure Size 83B
Color White
Diameter (Metric) Inner Neck 64 mm
Diameter (Metric) Outer 250 mm
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Height (Metric) With Closure 389 mm
Includes Handles
Material LDPE
No. per Case 6/Cs.
No. per Pack 1 Ea.
Product Type Carboy with Handle
Shape Round
Sterility Non-sterile
Workflow Step Upstream process solution storage. Upstream Validation
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.