missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure

Accurately fill and pour with these narrow-mouth, LDPE low particulate/low metals bottles with closure.

Supplier:  Thermo Scientific™ 3820030008

 View more versions of this product

Catalog No. 03-313-101


Only null left

Description

Description

Accurately pour and fill Thermo Scientific™ Nalgene™ LDPE Low Particulate/Low Metals Bottles with Closure, which feature a narrow mouth. These low-density polyethylene bottles include certification to contain less than 20 particles per mL at 0.3 microns or greater.

Metals certified to microliter (ppb) levels <0.20Hg, <0.5 Be, <1.0 As, Cd, Pb, <2.0 Sb,, Se, <5.0 Ag, Co, Cr, Cu, Mn, Tl, V, <10 Ba, Ni, Zn, <75 Al, <100 Ca, Fe, K, Na. Each bottle is double bagged under Class 10 laminar flow hoods inside a Class 100 clean room. Excellent for ICP-MS reagent and standard storage. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with specifications.
  • Certified to particulate and trace metals levels
  • Linerless polypropylene closures provide the ultimate in leakproof protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents
  • Exhibits flexibility, impact resistance and excellent visibility of contents for a wide variety of uses in the lab
  • Excellent chemical resistance to most acids, bases and alcohols make this a versatile bottle
  • Translucent
  • Autoclavable/Nonsterile
  • Guaranteed leakproof†
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications

Specifications

8 oz., 250 mL
8 oz.
Low Particulate/Low Metals Bottle
LDPE
1
Natural, translucent
24 mm
Yes
Round
Low Particulate/Low Metals Bottle
5.23 in.
Polypropylene
61 mm
No
250 mL
Translucent
18 mm
72
Non-sterile
Yes
Certified to particulate and trace metals levels
Narrow
133 mm
2.4 in.
Unlined
24 mm
Screw Cap
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.