missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure

Store corrosive chemicals and distilled water with these large, narrow-mouth PPCO bottles, which are resistant to most acids, bases and alcohols.

Supplier:  Thermo Scientific™ 22030010

 View more versions of this product

Catalog No. 02-924B



Only null left

Description

Description

Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure, which are resistant to most acids, bases and alcohols. Large sizes of these polypropylene copolymer bottles have a built-in shoulder loop.

Features:
  • 4 and 8L feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Lighter weight than glass alternatives make for easy lifting
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents
  • Autoclavable/Leakproof†

Includes:

Polypropylene screw caps

Note:

Completely disengage threads or remove closure before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Specifications

Specifications

1 gal., 4 L
1 gal.
Large Narrow-Mouth Bottle
PPCO
1
White
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Narrow
333 mm
13.1 in.
Polypropylene
152 mm
No
4 L
Translucent
25 mm
6
Non-sterile
38 mm
Yes
Round
Narrow Mouth Bottle
Unlined
38-430 mm
Screw Cap
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.