missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Large HDPE Funnels

Description
Replace glass and metal lab funnels with Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are both shatterproof and dentproof making them a valuble asset to lab safety programs.
- High-density polyethylene is shatterproof, making it a reliable long-term tool for the lab
Specifications
Specifications
| Diameter (Metric)Top | 201mm |
| Capacity (Metric) | 1.9 L |
| Diameter (Metric) Stem | 27 mm |
| Material | HDPE |
| Height (Metric) | 246 mm |
| Autoclavable | Not Autoclavable |
| Description | Funnel Large HDPE 203mm |
| Length (Metric) Stem | 88 mm |
| Quantity | 1/Pk.; 6/Cs. |