missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Large HDPE Funnels
GREENER_CHOICE

Catalog No. 103715A
Change view
Click to view available options
Diameter (Metric)Top:
201mm
258mm
Capacity (Metric):
1.9 L
4.06 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top Capacity (Metric)
103715A 201mm 1.9 L
103715B 258mm 4.06 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 103715A Supplier Thermo Scientific™ Supplier No. 42620080
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose a safer alternative to glass with this shatterproof funnel, featuring external and internal ribbing for air release.

Replace glass and metal lab funnels with Thermo Scientific™ Nalgene™ Large HDPE Funnel, featuring external and internal ribbing for air release. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are both shatterproof and dentproof making them a valuble asset to lab safety programs.

  • High-density polyethylene is shatterproof, making it a reliable long-term tool for the lab

Specifications

Diameter (Metric)Top 201mm
Capacity (Metric) 1.9 L
Diameter (Metric) Stem 27 mm
Material HDPE
Height (Metric) 246 mm
Autoclavable Not Autoclavable
Description Funnel Large HDPE 203mm
Length (Metric) Stem 88 mm
Quantity 1/Pk.; 6/Cs.