Learn More
Thermo Scientific™ Nalgene™ L900 Liquid Detergent
GREENER_CHOICE
Catalog No. 04319B
Change view
Click to view available options
Volume (Metric):
4 L
Catalog No. Volume (Metric)
04319B 4 L
1 options
Catalog No. 04319B Supplier Thermo Scientific™ Supplier No. 9004000
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thoroughly clean all plastic labware, with an environmentally friendly formula that will not cause crazing, stress cracking or clouding when used as directed.
Thoroughly clean all plastic labware, especially polycarbonate, with Thermo Scientific™ Nalgene™ L900 Liquid Detergent. This environmentally friendly formula will not cause crazing, stress cracking or clouding of plastic when used as directed.
  • For hand or machine use
  • Biodegradable
  • Non-toxic
  • Low foaming
  • Specially formulated for Nalgene plastic labware
Specifications
Description L900 Liquid Detergent; L-900 Liquid Detergent *DO NOT FREEZE* 4 Liter
Quantity 4 Pk.
Volume (English) 1.06 gal.
Volume (Metric) 4 L