missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE, Heavy-Duty Rectangular Carboy
GREENER_CHOICE

Catalog No. 0296156a
Change view
Click to view available options
Quantity:
Case of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
0296156A Case of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0296156A Supplier Thermo Scientific™ Supplier No. 22140050
Only null left

For Use With: Store and transport reagents.; Color: White

Store and transport reagents with the rugged Thermo Scientific™ Nalgene™ Heavy-Duty HDPE Carboy. The rectangular shape helps optimize limited storage space, and the large handle is easy to use, even with gloved hands. This heavy-duty polyethelyne carboy is both durable and space-saving.

Features:

  • Heavy walled construction for durability and chemical resistance
  • Graduated to 5 L and 1 gal. increments
  • Valve-sealing graduated/leakproof† linerless 70 mm screw closure
  • Not Autoclavable

Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications

Autoclavable Not Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Liner Unlined
Closure Size 70 mm
Closure Type Screw
Color White
Dimensions (L x W x H) 12.1 x 9.2 x 15.7 in. (323 x 234 x 399 mm)
For Use With (Application) Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Material HDPE
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Rectangular Carboy
Sterility Sterile
Quantity Case of 4
Workflow Step Upstream process solution storage. Upstream Validation
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes.