Learn More
Thermo Scientific™ Nalgene™ Heavy-Duty LDPE Funnels
GREENER_CHOICE
Avoid breakage and corrosion with these heavy-duty funnels, the toughest large funnels available.
Supplier:  Thermo Scientific™ 42600060
 View more versions of this product
Catalog No. 10371BB
Only null left
Description
Description
Avoid breakage and corrosion with Thermo Scientific™ Nalgene™ Heavy-Duty LDPE Funnels, the toughest large funnels available. Lighter weight than glass or metal funnels, these low density polyethylene funnels provide you with long-term use and are a valuble asset to lab safety programs.
  • Durable large funnels are designed for heavy-duty use
  • Rigid construction, ideal for vacuum filtration, larger sizes have ribs and a tab
Specifications
Chemical Identifiers
LDPE
Not Autoclavable
Specifications
150 mm
18 mm
196 mm
Funnel Heavy-Duty LDPE 6 inch
1/Pk.; 12/Cs.
777 mL
LDPE
Not Autoclavable
76mm
Product Suggestions
Product Suggestions