Learn More
Thermo Scientific™ Nalgene™ HDPE Quick Disconnects
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ Quick Disconnects.
$221.29 - $307.07
Specifications
| Material | HDPE |
|---|---|
| Packaging Quantity | 12/Pk., 72/Cs. |
| Product Type | Quick Disconnect |
| Catalog Number | Mfr. No. | For Use With | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With | Price | Quantity & Availability | |||||
15315A
![]() |
Thermo Scientific™
61500010 |
1/4 to 5/16 in. I.D. |
Pack of 12 Each for $221.29
Case of 6 Pack for $1,327.74
|
|
|||||
15315B
![]() |
Thermo Scientific™
61500020 |
3/8 to 7/16 in. I.D. |
Pack of 12 Each for $307.07
Case of 6 Pack for $1,842.42
|
|
|||||
Description
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ HDPE Quick Disconnects, which allow tubulations to slide together snugly with a twist.
Two tapered tubulations fit tightly together with male/female connection, allowing for rapid joining or separation of apparatus lines.Warranty: 90 days
Specifications
| HDPE | |
| 12/Pk., 72/Cs. | |
| Quick Disconnect |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.