missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE, Jerrican with Tethered Polypropylene Closure
GREENER_CHOICE

Catalog No. 0296023
Change view
Click to view available options
Includes:
Jerry Can with double port
Single Port
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
0296023 Single Port
0296024 Jerry Can with double port
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0296023 Supplier Thermo Scientific™ Supplier No. 22430013

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Use With: Ideal as a reservoir in automated systems; Closure Material: Polypropylene; Color: White

Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure, featuring a space-saving design, molded-in handle and recessed grip at the bottom to provide better control when pouring. The double-port jerryican (Cat. No. 2243-9013) is ideal as a reservoir in automated systems. The 53B closure can be plumbed with input/output tubing while the 38 mm closure is used for refilling or emptying the reservoir.

Key Features:

  • Jerricans are ideal as a reservoir in automated systems like the PRISM™ 3700 DAN Analyzer
  • Molded-in graduations in 1 gal. and 5 L increments allow for ease in dispensing
  • Molded-in handle provides a sturdy grip when carrying heavy contents
  • Integral shoulder handle and molded bottom grip make lifting comfortable
  • Wide stance and low center of gravity add stability
  • Heavy rugged design with good chemical resistance allows for a wide variety of solutions
  • Space-saving rectangular shape permits efficient storage

Ordering Information:

  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • Custom tubing sets can be designed at the Thermo Scientific Custom Tubing Set Configurator, www.thermoscientific.com/tubingconfigurator

Specifications

Autoclavable Not Autoclavable
Capacity (English) 3.5 gal.
Capacity (Metric) 13 L
Certifications/Compliance USP Class VI Compliant, FDA 21 CFR 177.1520 Compliant
Closure Color White
Closure Material Polypropylene
Closure Size 53B
Closure Type Tethered
Color White
Diameter (English) Inner Neck 1.5 in.
Diameter (Metric) Inner Neck 38 mm
Dimensions (L x W x H) 11 x 7 x 15 in. (290 x 189 x 378 mm)
For Use With (Application) Storage, Transportation ​
Includes Single Port
Material HDPE
Mouth Style Narrow
No. per Case 4/Cs.
No. per Pack 1 Ea.
Product Type Jerry Can
Sterility Non-sterile
Workflow Step Upstream process solution storage. Upstream Validation
Show More Show Less