missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with these HDPE diagnostic bottles without closure.

Supplier:  Thermo Scientific™ 3220200125

 View more versions of this product

Catalog No. 02-923-155


Only null left
Explore available promotions

Description

Description

Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure. Safe and leakproof† when used with Nalgene closures, these bottles can also be used in test kits and many other small-volume applications. Bottles are made of opaque white, high-density polyethylene and are packed without closures. They can be matched with various colored closures (Cat. No. 362150 series).

Features:
  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof† when assembled with Nalgene closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. 2000 per case. Order colored closures separately (Cat. No. 362150).

Specifications

Specifications

0.125 oz., 4 mL
0.125 oz.
8 mm
2000
Non-sterile
Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition.
No
Round
Sample Container
1.61 in.
13-415 mm
4 mL
Opaque White
HDPE
2000
13-415 mm
No
Narrow
41 mm
0.62 in.
Polypropylene
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.