missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Griffin Low-Form Plastic Beaker Variety Pack
GREENER_CHOICE

Catalog No. 02540271
Change view
Click to view available options
Capacity (Metric):
50 mL, 100 mL, 150 mL, 250 mL, 400 mL, 600 mL, and 1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02540271 50 mL, 100 mL, 150 mL, 250 mL, 400 mL, 600 mL, and 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02540271 Supplier Thermo Scientific™ Supplier No. 12011234
Only null left

A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Seven sizes, from 50 to 1000mL.

A convenient and cost effective way to get started setting up a new lab, assembling student lab kits, or refreshing an existing lab. Excellent chemical resistance to a wide variety of lab reagents, easy to read silk screened graduations and convenient stacking capabilities make these beakers an excellent choice for daily lab activities. Thermo Scientific™ Nalgene™ Griffin Low-Form Beaker Variety Pack includes one each 50mL, 100mL, 150mL, 250mL, 400mL, 600mL and 1000mL PPCO beakers.

  • Includes seven beakers, one each size: 50mL, 100mL, 150mL, 250mL, 400mL, 600mL and 1000mL
  • No drip single-spout design provides precision pouring control
  • With ribs for easy stacking
  • Silk screened with graduations, size code, resin code, catalog number, max/ temperature and “No Flame” symbol for easy identification and safe use
  • Impact resistant at temperatures above -40°C
  • Autoclavable/Graduated
  • High density polyethylene

Compliance:
  • Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Beakers and 21 CFR Part 177.1520 for food/beverage use.

Specifications

Color Natural translucent
Description Griffin Low-Form Beaker Variety Pk.
Capacity (Metric) 50 mL, 100 mL, 150 mL, 250 mL, 400 mL, 600 mL, and 1000 mL
Material PPCO
Quantity 1/Cs.
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Certifications/Compliance ISO/DIN 7056, 21 CFR Part 177.15
Scale mL
Style Griffin, Low Form
ACT Label Yes
Show More Show Less

Notes:

  • Autoclaving will affect graduation accuracy.