missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CryoTube™ Holders
GREENER_CHOICE

Catalog No. 033377E
Change view
Click to view available options
Holds:
25 Cryogenic Tubes
40 Cryogenic Tubes
50 Cryogenic Tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
033377E 50 Cryogenic Tubes
033377Z 25 Cryogenic Tubes
12565178 40 Cryogenic Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 033377E Supplier Thermo Scientific™ Supplier No. 50300510

Increase the functionality of your cryogenic tubes with these benchtop tube and vial holders in a variety of compartment configurations with single-handed capping and decapping.

Expand the functionality of cryogenic tube products with Thermo Scientific™ CryoTube™ Holders — benchtop tube and vial containers for single-handed capping and decapping. The product range includes Nalgene™ and Nunc™ containers in various sizes and compartment configurations.

  • Polystyrene coders snap into the caps of Nalgene Cryogenic Tubes for easy sample identification
  • Polycarbonate Benchtop Cryogenic Tube Holder has anti-rotation feature for one handed capping and decapping

Specifications

Anti-Rotation Yes
Color White
Material Polycarbonate
Type Vial Holder
For Use With (Equipment) Cryo Tube
Holds 50 Cryogenic Tubes
Product Line Nalgene
Tube Array 5 x 10