Learn More
Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Obtain a comprehensive solution when plastic is preferred to fiberboard.
Supplier: Thermo Scientific™ CS509X24
Description
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
- Warranty varies by country, please contact us for details
Chemical Identifiers
| Cryogenic Storage Box |
Specifications
| Cryogenic Storage Box | |
| 1.5 mL | |
| 2 in. | |
| 1 and 1.5 mL Vials |
| 5.29 cu. ft. | |
| Locator Jr. Plus, Locator 4 Plus, Locator 6 Plus | |
| 100 | |
| 10/Cs. |