missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
GREENER_CHOICE

Catalog No. 0128843
Change view
Click to view available options
Closure Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
0128843 White
02925106 Yellow
02925108 Green
02925109 Red
02925107 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 0128843 Supplier Thermo Scientific™ Supplier No. 3421580021

Eliminate the need for a rubber stopper with the safe-sealing solution of these continuous thread lined closures for serum vials.

Eliminate the need for a rubber stopper with the safe-sealing solution of high density polyetylene (HDPE) Thermo Scientific™ Nalgene™ Continuous Thread Lined Closures for Serum Vials.

Features:

  • Sterile to 10-6 SAL
  • Liner made of a low density polyethylene (LDPE) core between two facings of solid HDPE
  • Multiple closure colors available

Ordering Information:

Serum vial closures are bulk packed at 2,000 per case.

Specifications

Closure Color White
Description Serum vial closures
Material (Liner) Three-ply co-extruded liner (LDPE core between two facing of solid HDPE)
Color White
Diameter (Metric) Outer 22.1 mm
Material HDPE
Sterility Sterile
No. per Case 2000
No. per Pack 2000
Product Line Nalgene
Closure Material Rubber
Closure Type Lined Continuous-thread Closure
Quantity 2000 Cs.
Closure Size 20-415
For Use With (Equipment) Sterile PETG Serum Vials (Sold Separately)
Certifications/Compliance ISO 13485:2016
Height (Metric) 14.2 mm
Product Type Serum Vial Closure
Show More Show Less

The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.