missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners

Description
Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.
- Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
- Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic
Specifications
Specifications
| Type | Tank Liner |
| Description | Gamma Irradiated: 5 gal. (19L) |
| Material | Polyethylene |
| Capacity (English) | 5 gal. |
| Capacity (Metric) | 19 L |
| For Use With (Equipment) | Nalgene cylindrical tanks from 5 gal (19L) to 200 |
| No. per Case | 10 |
| No. per Pack | 10 |
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.