Learn More
Thermo Scientific™ Nalgene™ Closures for Dropper Bottles: Bulk Pack
Order these closures, which are available in multiple colors to help identify reagents, to complete your dropper bottle system.
Supplier: Thermo Scientific™ 3127600010
Description
Order Thermo Scientific™ Nalgene™ Closures for Dropper Bottles to complete the Nalgene Dropper Bottle System. Closures fit dropper bottles Cat. Nos. 312750 and 31275, and are available in multiple colors to help identify reagents.
Features:- Polypropylene
- Available in various colors
Ordering Information:
Dropper Bottle Closures are bulk packed 2000 per case. Order Nalgene controlled-drop dispensing tip (Cat. Nos. 312758, 312759) and dropper bottles (Cat. Nos. 312750, 312751) separately.
Specifications
| White | |
| 2000 | |
| 15-415 | |
| Yes | |
| Dropper Bottles 312750 and 312751. |
| Polypropylene | |
| 2000 | |
| Manufactured in compliance with ISO 13485:2003 quality requirements. Dropper Bottle Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Dropping Bottle Screw Cap | |
| Polypropylene |