Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks

Description
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:
- Tanks may be fitted with spigots
- Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
Specifications
Specifications
| Type | Closed-dome Tank |
| Diameter (English) Outer | 18.5 in. |
| Certifications/Compliance | Comply with FDA Reg. 177.1520 and USP Class VI |
| Diameter (Metric) Outer | 47 cm |
| Material | Polypropylene |
| Capacity (English) | 20 gal. |
| Capacity (Metric) | 75 L |
| Height (English) | 35 in. |
| Height (Metric) | 81 cm |
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.