Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Bio Tank Closure with Mixer Support Assembly
GREENER_CHOICE
Catalog No. 14831311M
Change view
Click to view available options
Closure Size:
152.3 mm
Catalog No. Closure Size
14831311M 152.3 mm
1 options
Catalog No. 14831311M Supplier Thermo Scientific™ Supplier No. 26510200
Enable overhead mixing in a closed system with this unique flange assembly that can be used with all sizes of Thermo Scientific™ Nalgene™ Closed-Dome Tanks.
Enable overhead mixing in a closed system with this unique, sanitary flange assembly. Thermo Scientific™ Nalgene™ Closed-Dome Bio Tank Closure with Mixer Support Assembly is for use with all sizes of Nalgene™ Closed-Dome Bio Tanks. Nalgene™ Biotech Mixer is not included.
Features:
  • 6 in. (15.2cm) polypropylene screw closure with 2 in. (5.1cm) sanitary ferrule welded in the center
  • 2 in. (5.1cm) silicone gasket and PDVF True Union clamp
  • Autoclavable
Compatible with:
Use with Nalgene Closed-Dome Tanks (Series 2650)
Specifications
Type Bio Tank
Description Closed dome tank closure w/support assembly
Closure Size 152.3 mm
Material Polypropylene, PVDF, Silicone
No. per Case 1
No. per Pack 1
Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.