missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ BioTransport Carrier
GREENER_CHOICE

Catalog No. 152512
Change view
Click to view available options
Material:
Polycarbonate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
152512 Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.

Catalog No. 152512 Supplier Thermo Scientific™ Supplier No. 71350001

To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.

Easily view contents with the Thermo Scientific™ Nalgene™ BioTransport Carrier.

Transport racks containing tubes or sample holders filled with potentially infectious materials using the Thermo Scientific™ Nalgene™ BioTransport Carrier, which offers excellent visibility of contents.

  • Polycarbonate carrier is tough and break resistant
  • Easy to grasp, molded-in handles
  • Silicone gasket seals tightly
  • Clamps securely hold carrier closed and assure leak-resistant seal
  • Stainless-steel handle available separately (7136-0001)
  • For accidental spills inside of carrier, autoclave with clamps closed

To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.To avoid exposure to biohazardous material due to accidental spills within carrier, autoclave and dispose of carrier and contents without opening clamps.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications

Certifications/Compliance Meets OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
Description BioTransport Carrier
Autoclavable Autoclavable
Material Polycarbonate
No. per Case 4
No. per Pack 1
Dimensions (L x W x H) 14.5 x 7.3 x 6.687 in. (36.8 x 18.4 x 17.0cm)
Length (Metric) 36.8 cm
Length (English) 14.5 in.
Width (English) 7.3 in.
Height (English) 6.687 in.
Product Type BioTransport Carrier
Height (Metric) 17.0 cm
Width (Metric) 18.4 cm
Show More Show Less