missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Autoclavable Septum Closure, Autoclavable, 38-430 size

Description
Enable aseptic injection of reagent or sample withdrawal without compromising sterility or integrity of contents with Thermo Scientific™ Nalgene™ Autoclavable Septum Closure, suitable for use with any bottle or container with 38-430 neck, including Nalgene culture vessels.
- Use with 18-gauge or smaller needles
- Polypropylene closure with thermoplastic elastomer septum
Specifications
Specifications
| Color | White |
| Description | Autoclavable Septum Closures |
| Material | Polypropylene |
| No. per Case | 12 |
| Quantity | Each |
| Sterility | Non-sterile |
| Thread Size (Metric) | S2 mm |
| Neck Finish | 38-430 |
| Mouth | Threaded |
| For Use With (Equipment) | Nalgene bottles with 38-430 threaded mouth. |
| Show More |