missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ 4-in-1 EZ Tote™ Bottle Carrier
GREENER_CHOICE

Catalog No. 02923170
Change view
Click to view available options
Holds:
Bottles (500 mL), Bottles (1 L), Bottles (4 L)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
02923170 Bottles (500 mL), Bottles (1 L), Bottles (4 L)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02923170 Supplier Thermo Scientific™ Supplier No. 65650001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Transport bottles and other lab essentials safely and easily using the Thermo ScientificTM NalgeneTM 4-in-1 EZ ToteTM Bottle Carrier.

Transport bottles and other lab essentials safely and easily using the Thermo Scientific™ Nalgene™ 4-in-1 EZ Tote™ Bottle Carrier. Manufactured from 3mm corrugated HDPE plastic, it holds eight 500mL, six 1L, or two 4L bottles. Versatile, durable and lightweight, carriers include partitions and assembly instructions, printed on each tote.

  • Reconfigurable partitions make this a versatile tote
  • Holds up to 28 lb. (12.7kg)
  • Stores flat and easy to assemble
  • Note: Carrier is not autoclavable

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.

Specifications

Description 4-in-1 EZ Tote TM HDPE Blue 13L x 8W x 13.5H
Autoclavable Not Autoclavable
Color Blue
Holds Bottles (500 mL), Bottles (1 L), Bottles (4 L)
Material HDPE
No. per Case 4
No. per Pack 1
Product Type Bottle Carrier