missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naftopidil hydrochloride, 98+%, Thermo Scientific™
An alpha1-adrenoceptor antagonist
Chemical Identifiers
| CAS | 1164469-60-6 |
|---|---|
| Molecular Formula | C24H29ClN2O3 |
| Molecular Weight (g/mol) | 428.957 |
| MDL Number | MFCD09971016 |
| InChI Key | VQAAEWMEVIOHTJ-UHFFFAOYSA-N |
| Synonym | naftopidil hydrochloride, opera_id_518, 1-4-2-methoxyphenyl piperazin-1-yl-3-naphthalen-1-yloxypropan-2-ol hydrochloride, 4-2-methoxyphenyl-?-1-naphthalenyloxy methyl-1-piperazineethanol hydrochloride |
| PubChem CID | 6603044 |
| IUPAC Name | 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol;hydrochloride |
| SMILES | COC1=CC=CC=C1N2CCN(CC2)CC(COC3=CC=CC4=CC=CC=C43)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ63249MA
|
N/A | 10mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AAJ63249MF
|
N/A | 50mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1164469-60-6 | |
| 428.957 | |
| VQAAEWMEVIOHTJ-UHFFFAOYSA-N | |
| 6603044 | |
| COC1=CC=CC=C1N2CCN(CC2)CC(COC3=CC=CC4=CC=CC=C43)O.Cl |
| C24H29ClN2O3 | |
| MFCD09971016 | |
| naftopidil hydrochloride, opera_id_518, 1-4-2-methoxyphenyl piperazin-1-yl-3-naphthalen-1-yloxypropan-2-ol hydrochloride, 4-2-methoxyphenyl-?-1-naphthalenyloxy methyl-1-piperazineethanol hydrochloride | |
| 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol;hydrochloride |
Specifications
| 1164469-60-6 | |
| MFCD09971016 | |
| Soluble in DMSO; Also soluble in ethanol; Insoluble in water. | |
| COC1=CC=CC=C1N2CCN(CC2)CC(COC3=CC=CC4=CC=CC=C43)O.Cl | |
| 428.957 | |
| 428.96 | |
| Naftopidil hydrochloride |
| C24H29ClN2O3 | |
| naftopidil hydrochloride, opera_id_518, 1-4-2-methoxyphenyl piperazin-1-yl-3-naphthalen-1-yloxypropan-2-ol hydrochloride, 4-2-methoxyphenyl-?-1-naphthalenyloxy methyl-1-piperazineethanol hydrochloride | |
| VQAAEWMEVIOHTJ-UHFFFAOYSA-N | |
| 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-naphthalen-1-yloxypropan-2-ol;hydrochloride | |
| 6603044 | |
| ≥98% |
Safety and Handling
RTECSNumber : TL9336500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only