missing translation for 'onlineSavingsMsg'
Learn More

Nadifloxacin 98.0+%, TCI America™

Catalog No. N0931200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0931200MG 200 mg
N09311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0931200MG Supplier TCI America Supplier No. N0931200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124858-35-1
Molecular Formula C19H21FN2O4
Molecular Weight (g/mol) 360.385
MDL Number MFCD00865081
InChI Key JYJTVFIEFKZWCJ-UHFFFAOYSA-N
Synonym 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid
PubChem CID 4410
ChEBI CHEBI:31889
SMILES CC1CCC2=C3N1C=C(C(=O)C3=CC(=C2N4CCC(CC4)O)F)C(=O)O

Specifications

Melting Point 247°C
Color White-Yellow
Quantity 200 mg
Formula Weight 360.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nadifloxacin
TSCA No
Recommended Storage Refrigerator