missing translation for 'onlineSavingsMsg'
Learn More

Nabumetone

Catalog No. AAJ6307206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6307206 5 g
AAJ6307214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6307206 Supplier Thermo Scientific Chemicals Supplier No. J6307206
Only null left

CAS: 42924-53-8 | C15H16O2 | 228.291 g/mol

Anti-inflammatory, anti-bacterial. A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Anti-inflammatory, anti-bacterial
  • A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase

Chemical Identifiers

CAS 42924-53-8
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.291
MDL Number MFCD00079518
InChI Key BLXXJMDCKKHMKV-UHFFFAOYSA-N
Synonym nabumetone, relafen, 4-6-methoxynaphthalen-2-yl butan-2-one, arthaxan, nabumetona, relifex, listran, relifen, nabumeton, consolan
PubChem CID 4409
ChEBI CHEBI:7443
IUPAC Name 4-(6-methoxynaphthalen-2-yl)butan-2-one
SMILES CC(=O)CCC1=CC2=C(C=C1)C=C(C=C2)OC

Specifications

Melting Point 76°C to 78°C
Color White
Quantity 5 g
Merck Index 14,6343
Solubility Information Soluble in alcohol or chloroform
Formula Weight 228.29
Physical Form Crystalline Powder
Chemical Name or Material Nabumetone
RTECSNumber EL9085000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only