Learn More
3M™ N95 Particulate Respirator with Nuisance Level Organic Vapor Relief†
Catalog No. 19034803
Change view
Click to view available options
With Faceseal:
Yes
Catalog No. With Faceseal
19034803 Yes
1 options
Catalog No. 19034803 Supplier 3M™ Supplier No. 7000002083
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NIOSH approved respirator used for nuisance level organic vapors such as solvents, degreasers, and/or resins.
  • NIOSH approved N95 rating
  • Cake resistant filter media
  • Foam faceseal
  • Adjustable buckle straps
  • Flame resistant per modified ASTM D2859
  • Cool Flow™ Exhalation Valve
  • Carbon layer for ozone/nuisance level organic vapors†
  • † Nuisance level refers to concentrations not exceeding OSHA PEL or applicable occupational exposure limit whichever is lower
Specifications
Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications/Compliance ASTM D2859
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-0454
Certifications (OSHA) OSHA 1910.134
Color Code White
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Polypropylene, Polyester
Product Type Particulate Respirator with Nuisance Organic Vapor Relief
Size Standard
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Assembly and mechanical, cleaning, composite finishing, foundry operations, grinding, paint preparation, petrochemical, processing of minerals, welding
Latex Free Yes
Material Headstrap Braided Polyisoprene, Cotton/Polyester
Model 8214
Nose Piece Material Aluminum
Nosepiece Type Standard
Nuisance Odor Relief Organic Vapors
Quantity 80/Cs.
Sterility Non-sterile
With Faceseal Yes
Strap Attachment Type Adjustable Buckle Straps
FDA Cleared Surgical Mask No
Show More Show Less