Learn More
Poltex N95 Mask Dispenser
Description
- Made with extremely durable, break resistant 3/16 in. thick PETG.
- Hinged Lid to keep dust out.
- Holds a stack of masks or the whole box.
- Hardware not included for wall mount unit (-W).
- Customer must provide their own screw/anchor for their wall type.
Spécifications
Spécifications
| Product Type | N95 Mask Dispenser |
| Material | PETG |
| For Use With (Application) | Healthcare |
| For Use With (Equipment) | N95 masks |
| Mounting | Wall Mount |