missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N719 Dye 75.0+%, TCI America™
Specifications
Specifications
| Melting Point | 260°C |
| Color | Black |
| Quantity | 100 mg |
| Formula Weight | 1188.56 |
| Percent Purity | ≥75.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(tetrabutylammonium) Dihydrogen Bis(isothiocyanato)bis(2,2′-bipyridyl-4,4′-dicarboxyl |
Safety and Handling
| TSCA | No |