missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Benzyladenine 99.0+%, TCI America™
$111.84 - $305.70
Chemical Identifiers
| CAS | 1214-39-7 |
|---|---|
| Molecular Formula | C12H11N5 |
| Molecular Weight (g/mol) | 225.26 |
| MDL Number | MFCD00005572 |
| InChI Key | NWBJYWHLCVSVIJ-UHFFFAOYSA-N |
| Synonym | 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap |
| PubChem CID | 62389 |
| ChEBI | CHEBI:29022 |
| IUPAC Name | N-benzyl-7H-purin-6-amine |
| SMILES | C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1 |
Chemical Identifiers
| 1214-39-7 | |
| 225.26 | |
| NWBJYWHLCVSVIJ-UHFFFAOYSA-N | |
| 62389 | |
| N-benzyl-7H-purin-6-amine |
| C12H11N5 | |
| MFCD00005572 | |
| 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap | |
| CHEBI:29022 | |
| C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1 |
Specifications
| 1214-39-7 | |
| White | |
| MFCD00005572 | |
| 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap | |
| C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1 | |
| 225.26 | |
| CHEBI:29022 | |
| ≥99.0% (HPLC,T) | |
| N6-Benzyladenine |
| 234°C | |
| C12H11N5 | |
| 5 g | |
| NWBJYWHLCVSVIJ-UHFFFAOYSA-N | |
| N-benzyl-7H-purin-6-amine | |
| 62389 | |
| 225.26 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-1137
RTECSNumber : AU6252200
TSCA : Yes