missing translation for 'onlineSavingsMsg'
Learn More

N6-Benzoyl-2'-deoxyadenosine Hydrate 98.0+%, TCI America™

Catalog No. p-7119950
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3101100MG 100 mg
B31011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3101100MG Supplier TCI America Supplier No. B3101100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 206752-42-3
Molecular Formula C17H19N5O5
Molecular Weight (g/mol) 373.369
MDL Number MFCD00149575
InChI Key WRMYMWRRSSDKIJ-LUHWTZLKSA-N
PubChem CID 16211731
IUPAC Name N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide;hydrate
SMILES C1C(C(OC1N2C=NC3=C2N=CN=C3NC(=O)C4=CC=CC=C4)CO)O.O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 355.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N6-Benzoyl-2′-deoxyadenosine Hydrate