missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-Benzoyl-2'-deoxyadenosine Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 355.35 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N6-Benzoyl-2′-deoxyadenosine Hydrate |
Safety and Handling
| TSCA | No |