missing translation for 'onlineSavingsMsg'
Learn More

N4-Benzoylcytosine 97.0+%, TCI America™

Catalog No. B31695G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31695G Supplier TCI America Supplier No. B31695G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26661-13-2
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD00239434
InChI Key XBDUZBHKKUFFRH-UHFFFAOYSA-N
Synonym n4-benzoylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl benzamide, n-benzoylcytosine, n-4-benzoyl cytosine, benzamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n-2-oxo-1h-pyrimidin-6-yl benzamide, benzamide, n-2,3-dihydro-2-oxo-4-pyrimidinyl, n-2-oxohydropyrimidin-4-yl benzamide, n-2-oxo-1h-pyrimidin-4-yl benzamide, n-benzoyl cytosine
PubChem CID 309343
IUPAC Name N-(2-oxo-2,3-dihydropyrimidin-4-yl)benzamide
SMILES O=C(NC1=CC=NC(=O)N1)C1=CC=CC=C1

Specifications

Melting Point 339°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.21
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N4-Benzoylcytosine