missing translation for 'onlineSavingsMsg'
Learn More

N4-Benzoylcytosine, 98%

Catalog No. AC460540050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460540050 5 g
AC460540250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460540050 Supplier Thermo Scientific Chemicals Supplier No. 460540050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 26661-13-2 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26661-13-2
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD00239434
InChI Key XBDUZBHKKUFFRH-UHFFFAOYSA-N
Synonym n4-benzoylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl benzamide, n-benzoylcytosine, n-4-benzoyl cytosine, benzamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n-2-oxo-1h-pyrimidin-6-yl benzamide, benzamide, n-2,3-dihydro-2-oxo-4-pyrimidinyl, n-2-oxohydropyrimidin-4-yl benzamide, n-2-oxo-1h-pyrimidin-4-yl benzamide, n-benzoyl cytosine
PubChem CID 309343
IUPAC Name N-(2-oxo-1H-pyrimidin-6-yl)benzamide
SMILES O=C(NC1=CC=NC(=O)N1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Elemental Analysis 59.86 to max. 62.93% min. Carbon 19.04 to max. 20.01% min. Nitrogen
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 215.21
Percent Purity 98%
Chemical Name or Material N4-Benzoylcytosine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only