missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Acetylcytosine 98.0+%, TCI America™
$50.33 - $167.76
Chemical Identifiers
| CAS | 14631-20-0 |
|---|---|
| Molecular Formula | C6H7N3O2 |
| Molecular Weight (g/mol) | 153.14 |
| MDL Number | MFCD00134466 |
| InChI Key | IJCKBIINTQEGLY-UHFFFAOYSA-N |
| Synonym | n4-acetylcytosine, n-acetylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide, acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n 4-acetylcytosine, n-2-oxo-1h-pyrimidin-6-yl acetamide, n-2-hydroxypyrimidin-4-yl acetamide, n-2-oxo-3h-pyrimidin-4-yl acetamide, 4-acetylamino-1,2-dihydro-2-pyrimidone |
| PubChem CID | 99309 |
| IUPAC Name | N-(2-oxo-2,3-dihydropyrimidin-4-yl)acetamide |
| SMILES | CC(=O)NC1=CC=NC(=O)N1 |
Chemical Identifiers
| 14631-20-0 | |
| 153.14 | |
| IJCKBIINTQEGLY-UHFFFAOYSA-N | |
| 99309 | |
| CC(=O)NC1=CC=NC(=O)N1 |
| C6H7N3O2 | |
| MFCD00134466 | |
| n4-acetylcytosine, n-acetylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide, acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n 4-acetylcytosine, n-2-oxo-1h-pyrimidin-6-yl acetamide, n-2-hydroxypyrimidin-4-yl acetamide, n-2-oxo-3h-pyrimidin-4-yl acetamide, 4-acetylamino-1,2-dihydro-2-pyrimidone | |
| N-(2-oxo-2,3-dihydropyrimidin-4-yl)acetamide |
Specifications
| 14631-20-0 | |
| White | |
| MFCD00134466 | |
| n4-acetylcytosine, n-acetylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide, acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n 4-acetylcytosine, n-2-oxo-1h-pyrimidin-6-yl acetamide, n-2-hydroxypyrimidin-4-yl acetamide, n-2-oxo-3h-pyrimidin-4-yl acetamide, 4-acetylamino-1,2-dihydro-2-pyrimidone | |
| CC(=O)NC1=CC=NC(=O)N1 | |
| 153.14 | |
| 153.14 | |
| Crystalline Powder |
| 325°C | |
| C6H7N3O2 | |
| 5 g | |
| IJCKBIINTQEGLY-UHFFFAOYSA-N | |
| N-(2-oxo-2,3-dihydropyrimidin-4-yl)acetamide | |
| 99309 | |
| ≥98.0% (T) | |
| N4-Acetylcytosine |
Safety and Handling
TSCA : Yes