missing translation for 'onlineSavingsMsg'
Learn More

N4-(3-Bromophenyl)quinazoline-4,6-diamine 98.0+%, TCI America™

Catalog No. B5416100MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5416100MG 100 mg
B541625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5416100MG Supplier TCI America Supplier No. B5416100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 169205-78-1
Molecular Formula C14H11BrN4
Molecular Weight (g/mol) 315.17
MDL Number MFCD09908082
InChI Key IZQHULBHKPGOAP-UHFFFAOYSA-N
PubChem CID 5328042
IUPAC Name N4-(3-bromophenyl)quinazoline-4,6-diamine
SMILES NC1=CC2=C(NC3=CC(Br)=CC=C3)N=CN=C2C=C1

Specifications

Melting Point 204°C
Color Green-Yellow
Quantity 100 mg
Formula Weight 315.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N4-(3-Bromophenyl)quinazoline-4,6-diamine
TSCA No
Recommended Storage Refrigerator