missing translation for 'onlineSavingsMsg'
Learn More

N2-Isobutyryl-2'-deoxyguanosine 98.0+%, TCI America™

Catalog No. I0700100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0700100MG 100 mg
I07001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0700100MG Supplier TCI America Supplier No. I0700100MG
Only null left

Chemical Identifiers

CAS 68892-42-2
Molecular Formula C14H19N5O5
Molecular Weight (g/mol) 337.336
MDL Number MFCD00010060
InChI Key SIDXEQFMTMICKG-DJLDLDEBSA-N
PubChem CID 688078
IUPAC Name N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
SMILES CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO)O

Specifications

Melting Point 300°C
Color White
Quantity 100 mg
Formula Weight 337.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N2-Isobutyryl-2′-deoxyguanosine
TSCA No
Recommended Storage Freezer