missing translation for 'onlineSavingsMsg'
Learn More

N1,N12-Diacetylspermine Hydrochloride Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022801
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022801 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022801 Supplier Merck Emd Millipore Supplier No. 91423100MG
Only null left

Specifications

CAS 61345-83-3 (Free Base)
Molecular Formula C14H30N4O2 · xHCl
Linear Formula C14H30N4O2 · xHCl
Quantity 100 mg
MDL Number MFCD30533822
UN Number NONH for all modes of transport
Synonym N,N'-[1,4-Butanediylbis(imino-3,1-propanediyl)]bis[acetamide] hydrochloride; DAS; N1,N12-Bis(acetyl)spermine hydrochloride
Formula Weight 286.41 (Free Base Basis)
Percent Purity ≥95% (HPLC)