missing translation for 'onlineSavingsMsg'
Learn More

N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine, 98%

Catalog No. AAH50291MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50291MD 250 mg
AAH5029103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50291MD Supplier Thermo Scientific Chemicals Supplier No. H50291MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6342-21-8 | C10H16N2 | 164.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6342-21-8
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD01631934
InChI Key NFSAPTWLWWYADB-UHFFFAOYSA-N
Synonym 2-amino-1-phenylethyl dimethylamine, n1,n1-dimethyl-1-phenyl-1,2-ethanediamine, n1,n1-dimethyl-1-phenylethane-1,2-diamine, n*1*,n*1*-dimethyl-1-phenyl-ethane-1,2-diamine, n∼1∼,n∼1∼-dimethyl-1-phenylethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-1-phenyl, acmc-20apq4, daa-0-0, n,n-dimethyl-1-phenyl-ethane-1,2-diamine
PubChem CID 241652
IUPAC Name N,N-dimethyl-1-phenylethane-1,2-diamine
SMILES CN(C)C(CN)C1=CC=CC=C1

Specifications

Quantity 250 mg
UN Number UN2735
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only