missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Aminophenyl)-N1-methylacetamide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 119-63-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
InChI Key QFELUFGHFLYZEZ-UHFFFAOYSA-N
Synonym n-4-aminophenyl-n-methylacetamide, 4'-amino-n-methylacetanilide, p-amino-n-methylacetanilide, acetamide, n-4-aminophenyl-n-methyl, n1-4-aminophenyl-n1-methylacetamide, 4-amino-n-acetyl-n-methylaniline, n-4-aminophenyl-n-methyl-acetamide, p-amino-n-acetyl-n-methylaniline, acetanilide, 4'-amino-n-methyl, 4-amino-n-methyl acetanilide
PubChem CID 67068
IUPAC Name N-(4-aminophenyl)-N-methylacetamide
SMILES CC(=O)N(C)C1=CC=C(C=C1)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BTB13402CB
View Documents
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB13402DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB13402DE
View Documents
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

119-63-1
164.208
n-4-aminophenyl-n-methylacetamide, 4'-amino-n-methylacetanilide, p-amino-n-methylacetanilide, acetamide, n-4-aminophenyl-n-methyl, n1-4-aminophenyl-n1-methylacetamide, 4-amino-n-acetyl-n-methylaniline, n-4-aminophenyl-n-methyl-acetamide, p-amino-n-acetyl-n-methylaniline, acetanilide, 4'-amino-n-methyl, 4-amino-n-methyl acetanilide
N-(4-aminophenyl)-N-methylacetamide
C9H12N2O
QFELUFGHFLYZEZ-UHFFFAOYSA-N
67068
CC(=O)N(C)C1=CC=C(C=C1)N

Specifications

119-63-1
Amber Glass Bottle
n-4-aminophenyl-n-methylacetamide, 4'-amino-n-methylacetanilide, p-amino-n-methylacetanilide, acetamide, n-4-aminophenyl-n-methyl, n1-4-aminophenyl-n1-methylacetamide, 4-amino-n-acetyl-n-methylaniline, n-4-aminophenyl-n-methyl-acetamide, p-amino-n-acetyl-n-methylaniline, acetanilide, 4'-amino-n-methyl, 4-amino-n-methyl acetanilide
CC(=O)N(C)C1=CC=C(C=C1)N
164.208
164.21
Technical
97%
C9H12N2O
QFELUFGHFLYZEZ-UHFFFAOYSA-N
N-(4-aminophenyl)-N-methylacetamide
67068
Tech..
N1-(4-Aminophenyl)-N1-methylacetamide
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents