missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Aminophenyl)-N1-methylacetamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB13402DE

Quantity 5g

 View more versions of this product

Catalog No. BTB13402DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

119-63-1
164.208
n-4-aminophenyl-n-methylacetamide, 4'-amino-n-methylacetanilide, p-amino-n-methylacetanilide, acetamide, n-4-aminophenyl-n-methyl, n1-4-aminophenyl-n1-methylacetamide, 4-amino-n-acetyl-n-methylaniline, n-4-aminophenyl-n-methyl-acetamide, p-amino-n-acetyl-n-methylaniline, acetanilide, 4'-amino-n-methyl, 4-amino-n-methyl acetanilide
N-(4-aminophenyl)-N-methylacetamide
C9H12N2O
QFELUFGHFLYZEZ-UHFFFAOYSA-N
67068
CC(=O)N(C)C1=CC=C(C=C1)N

Specifications

N1-(4-Aminophenyl)-N1-methylacetamide
97%
C9H12N2O
n-4-aminophenyl-n-methylacetamide, 4'-amino-n-methylacetanilide, p-amino-n-methylacetanilide, acetamide, n-4-aminophenyl-n-methyl, n1-4-aminophenyl-n1-methylacetamide, 4-amino-n-acetyl-n-methylaniline, n-4-aminophenyl-n-methyl-acetamide, p-amino-n-acetyl-n-methylaniline, acetanilide, 4'-amino-n-methyl, 4-amino-n-methyl acetanilide
CC(=O)N(C)C1=CC=C(C=C1)N
164.208
164.21
Technical
119-63-1
Amber Glass Bottle
5g
QFELUFGHFLYZEZ-UHFFFAOYSA-N
N-(4-aminophenyl)-N-methylacetamide
67068
Tech..
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents