missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-acetyl-3-hydroxyphenyl)acetamide, Thermo Scientific™

Catalog No. BTB10058DA
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB10058DA 1g
BTB10058DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB10058DA Supplier Thermo Scientific Chemicals Supplier No. BTB10058DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40547-58-8
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
InChI Key ZSSHNMZQDWSUJJ-UHFFFAOYSA-N
Synonym n-4-acetyl-3-hydroxyphenyl acetamide, n1-4-acetyl-3-hydroxyphenyl acetamide, 4'-acetamido-2'-hydroxyacetophenone, acetamide, n-4-acetyl-3-hydroxyphenyl, 4-acetamido-2-hydroxyacetophenone, n-4-acetyl-3-hydroxy-phenyl acetamide, n-4-acetyl-3-hydroxy-phenyl-acetamide
PubChem CID 2775185
IUPAC Name N-(4-acetyl-3-hydroxyphenyl)acetamide
SMILES CC(=O)C1=C(C=C(C=C1)NC(=O)C)O

Specifications

Quantity 1g
Formula Weight 193.2
Percent Purity Tech.
Grade Technical
Chemical Name or Material N1-(4-Acetyl-3-hydroxyphenyl)acetamide