missing translation for 'onlineSavingsMsg'
Learn More

N-Z-Ethylenediamine hydrochloride, 95%, Thermo Scientific™

Chemical Identifiers

CAS 18807-71-1
Molecular Formula C10H15ClN2O2
Molecular Weight (g/mol) 230.69
MDL Number MFCD00270150
InChI Key QMLKQXIAPAAIEJ-UHFFFAOYSA-N
Synonym benzyl n-2-aminoethyl carbamate hydrochloride, n-cbz-ethylenediamine hydrochloride, benzyl 2-aminoethyl carbamate hydrochloride, n-carbobenzoxy-1,2-diaminoethane hydrochloride, n-z-ethylenediamine hydrochloride, n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride, n-z-1,2-diaminoethane hydrochloride, n-cbz-1,2-diaminoethane hydrochloride, benzyl 2-aminoethyl carbamate hcl, n-benzyloxycarbonylethylenediamine hydrochloride
PubChem CID 12715871
IUPAC Name benzyl N-(2-aminoethyl)carbamate hydrochloride
SMILES Cl.NCCNC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC439810010
SDS View Documents
Thermo Scientific Chemicals
439810010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439810050
SDS View Documents
Thermo Scientific Chemicals
439810050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

18807-71-1
230.69
QMLKQXIAPAAIEJ-UHFFFAOYSA-N
12715871
Cl.NCCNC(=O)OCC1=CC=CC=C1
C10H15ClN2O2
MFCD00270150
benzyl n-2-aminoethyl carbamate hydrochloride, n-cbz-ethylenediamine hydrochloride, benzyl 2-aminoethyl carbamate hydrochloride, n-carbobenzoxy-1,2-diaminoethane hydrochloride, n-z-ethylenediamine hydrochloride, n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride, n-z-1,2-diaminoethane hydrochloride, n-cbz-1,2-diaminoethane hydrochloride, benzyl 2-aminoethyl carbamate hcl, n-benzyloxycarbonylethylenediamine hydrochloride
benzyl N-(2-aminoethyl)carbamate hydrochloride

Specifications

18807-71-1
100.0
94% min. (HPLC)
C10H15ClN2O2
1g
QMLKQXIAPAAIEJ-UHFFFAOYSA-N
benzyl N-(2-aminoethyl)carbamate hydrochloride
12715871
95%
94.0
Authentic
Glass bottle
MFCD00270150
benzyl n-2-aminoethyl carbamate hydrochloride, n-cbz-ethylenediamine hydrochloride, benzyl 2-aminoethyl carbamate hydrochloride, n-carbobenzoxy-1,2-diaminoethane hydrochloride, n-z-ethylenediamine hydrochloride, n-benzyloxycarbonyl-1,2-diaminoethane hydrochloride, n-z-1,2-diaminoethane hydrochloride, n-cbz-1,2-diaminoethane hydrochloride, benzyl 2-aminoethyl carbamate hcl, n-benzyloxycarbonylethylenediamine hydrochloride
Cl.NCCNC(=O)OCC1=CC=CC=C1
230.69
230.69
N-Z-Ethylenediamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only